1-Boc-4-bromopiperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS35469G1 AS35469G5
ACTIAS35469G1EA 0 GBP
ACTIAS35469G1 ACTIAS35469G5
1-Boc-4-bromopiperidine
1-Boc-4-bromopiperidine
Formula: C₁₀H₁₈BrNO₂
MDL Number: MFCD04115039
CAS Number: 180695-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR