1-(Phenylsulphonyl)pyrrolidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0483-5G
OT-0483-25G
OT-0483-1G
COBBOT-0483-5GEA
0
GBP
COBBOT-0483-5G
COBBOT-0483-25G
COBBOT-0483-1G
1-(Phenylsulphonyl)pyrrolidine
1-(Phenylsulphonyl)pyrrolidine
Formula:
C₁₀H₁₃NO₂S |
MDL Number:
MFCD01055979 CAS Number: 5033-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...