3-(Piperidin-1-ylsulphonyl)aniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0135-1G OT-0135-5G
COBBOT-0135-1GEA 0 GBP
COBBOT-0135-1G COBBOT-0135-5G
3-(Piperidin-1-ylsulphonyl)aniline
3-(Piperidin-1-ylsulphonyl)aniline
Formula: C₁₁H₁₆N₂O₂S
MDL Number: MFCD02063328
CAS Number: 22184-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR