4,4-Difluoropiperidine hydrochloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4-Difluoropiperidine HCl

AS13080G1 AS13080G5
ACTIAS13080G1EA 0 GBP
ACTIAS13080G1 ACTIAS13080G5
4,4-Difluoropiperidine hydrochloride
4,4-Difluoropiperidine hydrochloride
Formula: C₅H₁₀ClF₂N
MW: 157.59 g/mol
MDL Number: MFCD03095381
CAS Number: 144230-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR