(S)-(+)-3-Fluoropyrrolidine hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013157-1G 013157-500MG
FLUO013157-1GEA 0 GBP
FLUO013157-1G FLUO013157-500MG
(S)-(+)-3-Fluoropyrrolidine hydrochloride
(S)-(+)-3-Fluoropyrrolidine hydrochloride
Formula: C₄H₉ClFN
MDL Number: MFCD04038718
CAS Number: 136725-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR