1-(Dimethylcarbamoyl)piperidine-4-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23960G1
AS23960G25
AS23960G5
ACTIAS23960G1EA
231
GBP
ACTIAS23960G1
ACTIAS23960G25
ACTIAS23960G5
1-(Dimethylcarbamoyl)piperidine-4-carboxylic acid
1-(Dimethylcarbamoyl)piperidine-4-carboxylic acid
Formula:
C₉H₁₆N₂O₃ |
MDL Number:
MFCD03830325 CAS Number: 333985-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...