1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23957G1 AS23957G5
ACTIAS23957G1EA 106 GBP
ACTIAS23957G1 ACTIAS23957G5
1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
Formula: C₉H₁₆N₂O₃
MDL Number: MFCD03830326
CAS Number: 702670-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR