1-Boc-isonipecotic acid methyl ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate

AM-1023-25G AM-1023-100G AM-1023-5G
COBBAM-1023-25GEA 0 GBP
COBBAM-1023-25G COBBAM-1023-100G COBBAM-1023-5G
1-Boc-isonipecotic acid methyl ester
1-Boc-isonipecotic acid methyl ester
Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
MDL Number: MFCD02183584
CAS Number: 124443-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR