1-(3-Bromo-5-methylphenylsulphonyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6462-1G HC-6462-5G
COBBHC-6462-1GEA 0 GBP
COBBHC-6462-1G COBBHC-6462-5G
1-(3-Bromo-5-methylphenylsulphonyl)pyrrolidine
1-(3-Bromo-5-methylphenylsulphonyl)pyrrolidine
Formula: C₁₁H₁₄BrNO₂S
MDL Number: MFCD09972152
CAS Number: 1020252-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR