1-(3-Bromo-5-(trifluoromethyl)phenylsulphonyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-5-(pyrrolidin-1-ylsulphonyl)benzotrifluoride

HC-6309-5G HC-6309-1G
COBBHC-6309-5GEA 0 GBP
COBBHC-6309-5G COBBHC-6309-1G
1-(3-Bromo-5-(trifluoromethyl)phenylsulphonyl)pyrrolidine
1-(3-Bromo-5-(trifluoromethyl)phenylsulphonyl)pyrrolidine
Formula: C₁₁H₁₁BrF₃NO₂S
MW: 358.18 g/mol
MDL Number: MFCD09801012
CAS Number: 951884-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR