1-Furfurylpyrrole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Furan-2-ylmethyl)-1H-pyrrole

M-Clarity™ quality level = MQ300

W328405-100G-K W328405-1KG-K W328405-25G-K
SIALW328405-100G-KEA 885 GBP
SIALW328405-100G-K SIALW328405-1KG-K SIALW328405-25G-K
1-Furfurylpyrrole, Sigma-Aldrich®
1-Furfurylpyrrole
Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00010080
CAS Number: 1438-94-4
EINECS: 215-876-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR