Ethyl-4-hydroxy-2-piperidinopyrimidine-5-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2020-5G HC-2020-1G
COBBHC-2020-5GEA 0 GBP
COBBHC-2020-5G COBBHC-2020-1G
Ethyl-4-hydroxy-2-piperidinopyrimidine-5-carboxylate
Ethyl 4-hydroxy-2-piperidinopyrimidine-5-carboxylate
Formula: C₁₂H₁₆ClN₃O₂
MDL Number: MFCD00085757
CAS Number: 25693-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR