(5-Bromopyrimidin-2-yl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-(piperidin-1-yl)pyrimidine

PY-7999-5G PY-7999-25G
COBBPY-7999-5GEA 0 GBP
COBBPY-7999-5G COBBPY-7999-25G
(5-Bromopyrimidin-2-yl)piperidine
(5-Bromopyrimidin-2-yl)piperidine
Formula: C₉H₁₂BrN₃
MW: 242.1157 g/mol
MDL Number: MFCD00483257
CAS Number: 57356-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR