2-(1H-Pyrazol-4-yl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

016083-1G 016083-5G
FLUO016083-1GEA 0 GBP
FLUO016083-1G FLUO016083-5G
2-(1H-Pyrazol-4-yl)pyridine
2-(1H-Pyrazol-4-yl)pyridine
Formula: C₈H₇N₃
MW: 145.1613 g/mol
MDL Number: MFCD02579318
CAS Number: 439106-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR