3-Chloro-2-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2509-5G HC-2509-1G
COBBHC-2509-5GEA 0 GBP
COBBHC-2509-5G COBBHC-2509-1G
3-Chloro-2-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
3-Chloro-2-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
Formula: C₁₀H₇ClF₃N₃
MDL Number: MFCD12026021
CAS Number: 1150164-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR