3-Chloro-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2580-1G HC-2580-5G
COBBHC-2580-1GEA 0 GBP
COBBHC-2580-1G COBBHC-2580-5G
3-Chloro-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine
3-Chloro-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine
Formula: C₁₀H₁₀ClN₃
MDL Number: MFCD12026033
CAS Number: 1150164-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR