5-Bromo-2-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2508-1G
HC-2508-5G
COBBHC-2508-1GEA
0
GBP
COBBHC-2508-1G
COBBHC-2508-5G
5-Bromo-2-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
5-Bromo-2-(3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)pyridine
Formula:
C₁₀H₇BrF₃N₃ |
MDL Number:
MFCD12546464 CAS Number: 1187385-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...