2-(4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)-6-chloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2542-5G
HC-2542-1G
COBBHC-2542-5GEA
0
GBP
COBBHC-2542-5G
COBBHC-2542-1G
2-(4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)-6-chloropyridine
2-(4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl)-6-chloropyridine
Formula:
C₁₀H₆BrClF₃N₃ |
MDL Number:
MFCD12546466 CAS Number: 1187386-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...