2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-6-chloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2708-1G
HC-2708-5G
COBBHC-2708-1GEA
0
GBP
COBBHC-2708-1G
COBBHC-2708-5G
2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-6-chloropyridine
2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-6-chloropyridine
Formula:
C₁₀H₉BrClN₃ |
MDL Number:
MFCD12026043 CAS Number: 1150271-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...