2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-3-chloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2725-5G
HC-2725-1G
COBBHC-2725-5GEA
0
GBP
COBBHC-2725-5G
COBBHC-2725-1G
2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-3-chloropyridine
2-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-3-chloropyridine
Formula:
C₁₀H₉BrClN₃ |
MDL Number:
MFCD12026045 CAS Number: 1150271-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...