5-Bromo-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2581-5G HC-2581-1G
COBBHC-2581-5GEA 0 GBP
COBBHC-2581-5G COBBHC-2581-1G
5-Bromo-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine
5-Bromo-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine
Formula: C₁₀H₁₀BrN₃
MW: 252.1105 g/mol
MDL Number: MFCD12026034
CAS Number: 1150164-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR