Pilocarpine hydrochloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (+)-Pilocarpine hydrochloride

M-Clarity™ quality level = MQ100

P6503-10G P6503-50G P6503-5G
SIALP6503-10GEA 435 GBP
SIALP6503-10G SIALP6503-50G SIALP6503-5G
Pilocarpine hydrochloride, Sigma-Aldrich®
Pilocarpine hydrochloride
Formula: C₁₁H₁₇ClN₂O₂
MDL Number: MFCD00012722
CAS Number: 54-71-7
EINECS: 200-212-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR