4-(Tributylstannyl)-3,6-dihydro-2H-pyran

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51876G1 AS51876G5
ACTIAS51876G1EA 325 GBP
ACTIAS51876G1 ACTIAS51876G5
4-(Tributylstannyl)-3,6-dihydro-2H-pyran
4-(Tributylstannyl)-3,6-dihydro-2H-pyran
Formula: C₁₇H₃₄OSn
MDL Number: MFCD18325166
CAS Number: 535924-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR