3,5-Dimethyl-4-(tributylstannyl)isoxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51228G1 AS51228G5
ACTIAS51228G1EA 0 GBP
ACTIAS51228G1 ACTIAS51228G5
3,5-Dimethyl-4-(tributylstannyl)isoxazole
3,5-Dimethyl-4-(tributylstannyl)isoxazole
Formula: C₁₇H₃₃NOSn
MDL Number: MFCD01318973
CAS Number: 136295-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR