Ethyl-5-(tributylstannyl)isoxazole-3-carboxylate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

706981-1G 706981-500MG
SIAL706981-1GEA 556 GBP
SIAL706981-1G SIAL706981-500MG
Ethyl-5-(tributylstannyl)isoxazole-3-carboxylate, Sigma-Aldrich®
Ethyl 5-(tributylstannyl)isoxazole-3-carboxylate
Formula: C₁₈H₃₃NO₃Sn
MDL Number: MFCD07368612
CAS Number: 126085-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR