4-(5-Nitroquinolin-6-yl)cyclohexanol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2265-5G
HC-2265-1G
COBBHC-2265-5GEA
0
GBP
COBBHC-2265-5G
COBBHC-2265-1G
4-(5-Nitroquinolin-6-yl)cyclohexanol
4-(5-Nitroquinolin-6-yl)cyclohexanol
Formula:
C₁₅H₁₆N₂O₃ |
MDL Number:
MFCD12026011 CAS Number: 1150163-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...