5-Nitro-6-(piperidin-1-yl)quinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2430-5G
HC-2430-1G
COBBHC-2430-5GEA
0
GBP
COBBHC-2430-5G
COBBHC-2430-1G
5-Nitro-6-(piperidin-1-yl)quinoline
5-Nitro-6-(piperidin-1-yl)quinoline
Formula:
C₁₄H₁₅N₃O₂ |
MDL Number:
MFCD12026016 CAS Number: 19979-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...