4-(Piperidin-1-yl)-8-(trifluoromethyl)quinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6339-1G
HC-6339-5G
COBBHC-6339-1GEA
0
GBP
COBBHC-6339-1G
COBBHC-6339-5G
4-(Piperidin-1-yl)-8-(trifluoromethyl)quinoline
4-(Piperidin-1-yl)-8-(trifluoromethyl)quinoline
Formula:
C₁₅H₁₅F₃N₂ |
MDL Number:
MFCD09972138 CAS Number: 1020252-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...