1-(2,4-Difluorophenyl)biguanide hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF614.2.5 008288-1G 008288-5G
CHMPCFF614.2.5EA 0 GBP
CHMPCFF614.2.5 FLUO008288-1G FLUO008288-5G
1-(2,4-Difluorophenyl)biguanide hydrochloride
1-(2,4-Difluorophenyl)biguanide hydrochloride
Formula: C₈H₁₀ClF₂N₅
MDL Number: MFCD00053015
CAS Number: 66088-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR