1-(3-Chlorophenyl)biguanide hydrochloride ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B20355.03 B20355.06
B20355.03EA 19.6 GBP
B20355.03 B20355.06
1-(3-Chlorophenyl)biguanide hydrochloride ≥97%
1-(3-Chlorophenyl)biguanide hydrochloride

Purity: 97% CAS No.: 2113-05-5 Molecular Formula: C8H10ClN5.HCl Formula Weight: 248.12 Melting Point: 191-194° MDL No.: MFCD00053019

Formula: C₈H₁₁Cl₂N₅
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00053019
CAS Number: 2113-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR