4-(Morpholino)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS1486G1 AS1486G25 AS1486G5
ACTIAS1486G1EA 0 GBP
ACTIAS1486G1 ACTIAS1486G25 ACTIAS1486G5
4-(Morpholino)phenylboronic acid
4-(Morpholino)phenylboronic acid
Formula: C₁₀H₁₄BNO₃
MDL Number: MFCD03095169
CAS Number: 186498-02-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR