3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester, 3-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, tert-Butyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperazine-1-carboxylate

PN-8802-5G PN-8802-1G
COBBPN-8802-5GEA 0 GBP
COBBPN-8802-5G COBBPN-8802-1G
3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Formula: C₂₂H₃₃BN₂O₅
MW: 416.32 g/mol
MDL Number: MFCD09266185
CAS Number: 883738-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR