3-Chloro-4-(4-morpholinylcarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(morpholine-4-carbonyl)phenylboronic acid

BB-2769-5G BB-2769-1G
COBBBB-2769-5GEA 0 GBP
COBBBB-2769-5G COBBBB-2769-1G
3-Chloro-4-(4-morpholinylcarbonyl)phenylboronic acid
3-Chloro-4-(4-morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BClNO₄
MW: 253.49 g/mol
MDL Number: MFCD07363774
CAS Number: 850589-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR