4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, 4-(Morpholine-4-carbonyl)phenylboronic Acid Pinacol Ester

PN-3004-5G PN-3004-1G
COBBPN-3004-5GEA 0 GBP
COBBPN-3004-5G COBBPN-3004-1G
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₄BNO₄
MW: 317.19 g/mol
MDL Number: MFCD05863913
CAS Number: 656239-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR