3-(Morpholinomethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 3-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester

PN-5196-5G PN-5196-1G
COBBPN-5196-5GEA 0 GBP
COBBPN-5196-5G COBBPN-5196-1G
3-(Morpholinomethyl)phenylboronic acid pinacol ester
3-(Morpholinomethyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₃
MW: 303.20 g/mol
MDL Number: MFCD06797489
CAS Number: 364794-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR