(E)-3-(6-Fluoro-4-hydroxy-2-methylquinolin-3-yl)-N-(4-fluorophenyl)but-2-enamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0745-5G
OT-0745-1G
COBBOT-0745-5GEA
0
GBP
COBBOT-0745-5G
COBBOT-0745-1G
(E)-3-(6-Fluoro-4-hydroxy-2-methylquinolin-3-yl)-N-(4-fluorophenyl)but-2-enamide
(E)-3-(6-Fluoro-4-hydroxy-2-methylquinolin-3-yl)-N-(4-fluorophenyl)but-2-enamide
Formula:
C₂₀H₁₆F₂N₂O₂ |
MDL Number:
MFCD12026058 CAS Number: 1150271-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...