(E)-3-(4-Hydroxy-8-methoxy-2-methylquinolin-3-yl)-N-(2-methoxyphenyl)but-2-enamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0744-5G
OT-0744-1G
COBBOT-0744-5GEA
0
GBP
COBBOT-0744-5G
COBBOT-0744-1G
(E)-3-(4-Hydroxy-8-methoxy-2-methylquinolin-3-yl)-N-(2-methoxyphenyl)but-2-enamide
(E)-3-(4-Hydroxy-8-methoxy-2-methylquinolin-3-yl)-N-(2-methoxyphenyl)but-2-enamide
Formula:
C₂₂H₂₂N₂O₄ |
MDL Number:
MFCD11504952 CAS Number: 1072944-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...