2-Amino-4-(1H-pyrazol-1-yl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21549G1 AS21549G25 AS21549G5
ACTIAS21549G1EA 0 GBP
ACTIAS21549G1 ACTIAS21549G25 ACTIAS21549G5
2-Amino-4-(1H-pyrazol-1-yl)benzoic acid
2-Amino-4-(1H-pyrazol-1-yl)benzoic acid
Formula: C₁₀H₉N₃O₂
MDL Number: MFCD11656802
CAS Number: 1186663-55-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR