4-(1H-Pyrazol-1-yl)benzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20711G25
AS20711G5
ACTIAS20711G25EA
324
GBP
ACTIAS20711G25
ACTIAS20711G5
4-(1H-Pyrazol-1-yl)benzoic acid
4-(1H-Pyrazol-1-yl)benzoic acid
Formula:
C₁₀H₈N₂O₂ MW: 188.18 g/mol |
MDL Number:
MFCD02682056 CAS Number: 16209-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...