2-Thioxanthine ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B21606.03
B21606.06
B21606.03EA
61.4
GBP
B21606.03
B21606.06
2-Thioxanthine ≥98%
2-Thioxanthine
Purity: 98+%
CAS No.: 2487-40-3
Molecular Formula: C5H4N4OS
Formula Weight: 168.17
Melting Point:
Formula:
C₅H₄N₄OS Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031505 CAS Number: 2487-40-3 EINECS: 219-636-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...