Potassium(pyrrolidin-1-ylmethyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Potassium trifluoro[(pyrrolidin-1-yl)methyl]borate

TB-0021-1G TB-0021-5G
COBBTB-0021-1GEA 0 GBP
COBBTB-0021-1G COBBTB-0021-5G
Potassium(pyrrolidin-1-ylmethyl)trifluoroborate
Potassium (pyrrolidin-1-ylmethyl)trifluoroborate
Formula: C₅H₁₀BF₃KN
MDL Number: MFCD10700161
CAS Number: 888711-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR