5-Bromo-3-nitro-2-(piperidin-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7337-5G PY-7337-1G
COBBPY-7337-5GEA 0 GBP
COBBPY-7337-5G COBBPY-7337-1G
5-Bromo-3-nitro-2-(piperidin-1-yl)pyridine
5-Bromo-3-nitro-2-(piperidin-1-yl)pyridine
Formula: C₁₀H₁₂BrN₃O₂
MDL Number: MFCD10699707
CAS Number: 1033202-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR