3-Bromo-5-nitro-2-(piperidin-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7392-1G PY-7392-5G
COBBPY-7392-1GEA 0 GBP
COBBPY-7392-1G COBBPY-7392-5G
3-Bromo-5-nitro-2-(piperidin-1-yl)pyridine
3-Bromo-5-nitro-2-(piperidin-1-yl)pyridine
Formula: C₁₀H₁₂BrN₃O₂
MDL Number: MFCD11053787
CAS Number: 1065074-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR