2-Bromo-6-(piperidin-1-yl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6331-1G HC-6331-25G HC-6331-5G
COBBHC-6331-1GEA 0 GBP
COBBHC-6331-1G COBBHC-6331-25G COBBHC-6331-5G
2-Bromo-6-(piperidin-1-yl)pyridine
2-Bromo-6-(piperidin-1-yl)pyridine
Formula: C₁₀H₁₃BrN₂
MDL Number: MFCD07772823
CAS Number: 24255-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR