8-Chloro-2-methyl-4-quinazolone
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6225-5G
HC-6225-1G
COBBHC-6225-5GEA
0
GBP
COBBHC-6225-5G
COBBHC-6225-1G
8-Chloro-2-methyl-4-quinazolone
8-Chloro-2-methyl-4-quinazolone
Formula:
C₉H₇ClN₂O |
MDL Number:
MFCD09800989 CAS Number: 19407-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...