4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6480-1G HC-6480-5G
COBBHC-6480-1GEA 0 GBP
COBBHC-6480-1G COBBHC-6480-5G
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid
Formula: C₁₁H₆F₃NO₃
MDL Number: MFCD00047603
CAS Number: 23779-95-5
EINECS: 245-878-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR