4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20942G1 AS20942G25 AS20942G5
ACTIAS20942G1EA 0 GBP
ACTIAS20942G1 ACTIAS20942G25 ACTIAS20942G5
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid
Formula: C₁₁H₆F₃NO₃
MDL Number: MFCD00047603
CAS Number: 23779-95-5
EINECS: 245-878-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR