5-Bromo-N-isopropylpicolinamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Bromo-N-isopropylpyridine-2-carboxamide
PY-7197-1G
PY-7197-5G
COBBPY-7197-1GEA
0
GBP
COBBPY-7197-1G
COBBPY-7197-5G
5-Bromo-N-isopropylpicolinamide
5-Bromo-N-isopropylpicolinamide
Formula:
C₉H₁₁BrN₂O |
MDL Number:
MFCD09801058 CAS Number: 845305-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...