3-Benzyloxy-4-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5235-1G BB-5235-5G
COBBBB-5235-1GEA 0 GBP
COBBBB-5235-1G COBBBB-5235-5G
3-Benzyloxy-4-methoxyphenylboronic acid
3-Benzyloxy-4-methoxyphenylboronic acid
Formula: C₁₄H₁₅BO₄
MDL Number: MFCD10696652
CAS Number: 243990-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR