5-(Benzyloxy)-2-nitrotoluene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0727-5G OT-0727-25G
COBBOT-0727-5GEA 0 GBP
COBBOT-0727-5G COBBOT-0727-25G
5-(Benzyloxy)-2-nitrotoluene
5-(Benzyloxy)-2-nitrotoluene
Formula: C₁₄H₁₃NO₃
MDL Number: MFCD00024269
CAS Number: 22424-58-4
EINECS: 244-988-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR